missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entamoeba histolytica Monoclonal Antibody (EH34.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110655
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10655 detects Entamoeba histolytica. MA1-10655 has been successfully used in ELISA applications. The MA1-10655 immunogen is Entamoeba histolytica HK-9.
Entamoeba histolytica is an anaerobic parasitic amoebozoan, part of the genus Entamoeba. Predominantly infecting humans and other primates causing amoebiasis, E. histolytica is estimated to infect about 50 million people worldwide. E. histolytica infection is estimated to kill more than 55,000 people each year.Specifications
Entamoeba histolytica | |
Monoclonal | |
0.77 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG1 |
ELISA | |
EH34.5 | |
Unconjugated | |
E. histolytica | |
Entamoeba histolytica HK-9. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |