Learn More
3M™ Petrifilm™ Enterobacteriaceae Count Plates
Provide a cost-effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples and bottled water testing
Brand:  3M™ Petrifilm™ 7100126807
Encompass
Product Code. 17213784
  • £1042.86 / Pack of 1000
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Results in 24 hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
  • This plate has been awarded Official Methods of Analysis (OMA) #2003.01 for selected foods and NF VALIDATION certificate: 3M 01/6-09/97 for All human food products, Animal feed and Industrial environmental samples by AFNOR Certification.
  • Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Food and Beverage, Environmental Test
1000