missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Entinostat Product Code.: 17271282
compliance-icons compliance-icons

Thermo Scientific Chemicals Entinostat

Product Code. 17271282
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 17271282

Brand: Thermo Scientific Chemicals 466872500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209783-80-2
Molecular Formula C21H20N4O3

Specifications

Quantity 250 mg
Formula Weight 376.41
InChI Key INVTYAOGFAGBOE-UHFFFAOYSA-N
SMILES NC1=CC=CC=C1NC(=O)C1=CC=C(CNC(=O)OCC2=CC=CN=C2)C=C1
IUPAC Name (pyridin-3-yl)methyl N-({4-[(2-aminophenyl)carbamoyl]phenyl}methyl)carbamate
Molecular Weight (g/mol) 376.42
Chemical Name or Material Entinostat

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464968 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number466870010,466872500
Lot NumberA0451333 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number466870010,466872500
Lot NumberA0418872 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number466870010,466872500
compliance-icons compliance-icons
Product Identifier
  • Entinostat
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : N-(2-Aminophenyl)-4-[N-(pyridine-3-ylmethoxycarbonyl)aminomethyl]benzamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Entinostat >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.