Learn More
Sartorius Entris™ II Advanced Analytical Balances
GREENER_CHOICE
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
£2840.00 - £3875.00
Specifications
Certifications/Compliance CE Certificate, CSA for Canada
Connection Type D-sub connector, 9-pin; USB UTL; USB Device
Display Type Intuitive Graphic Touch Technology
Readability 0.1 mg
Material Glass, PBT, Stainless Steel
View More Specs
Products 7
Product Code Brand Capacity Calibration Price Quantity & Availability  
Product Code Brand Capacity Calibration Price Quantity & Availability  
16605693
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA64I-1S
60 g Internal
£2910.00
Each
16675683
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA64-1S
60 g
£3010.00
Each
16642454
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA124I-1S
120 g Internal
£3055.00
Each
16632454
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA124-1S
120 g
£2840.00
Each
16655653
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA224I-1S
220 g Internal
£3340.00
Each
16625653
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA224-1S
220 g
£3120.00
Each
16646253
GREENER_CHOICE
View accessory products View Documents
Sartorius
BCA324I-1S
320 g Internal
£3875.00
Each
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
CE Certificate, CSA for Canada
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤1.5 sec.
100/240 V
50/60 Hz
D-sub connector, 9-pin; USB UTL; USB Device
0.1 mg
Electronic Analytical Balance
Balance
0.2 A
Class I
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Entris™ II Advanced Analytical Balances