Learn More
Sartorius Entris™ II Precision Balance, Internal Adjustment
Entris™ II Precision Balance with Internal Adjustment offering isoCAL, LED touch technology, and 12 built-in applications at a budget price. Every balance offers quality, value and consistency.
£1685.00 - £2435.00
Specifications
Certifications/Compliance CEU
Connection Type RS232, USB C
Display Type Touchscreen
Material PBT, Glass, Stainless Steel
Type Precision Balance
View More Specs
Products 17
Product Code Brand Capacity Readability Model Price Quantity & Availability  
Product Code Brand Capacity Readability Model Price Quantity & Availability  
16354698
View accessory products View Documents
Sartorius
BCE223I-1CEU
220 g 1 mg CEU Electronic Toploading Balance
£1775.00
1 unit
16374708
View accessory products View Documents
Sartorius
BCE323I-1CEU
320 g 1 mg CEU Electronic Toploading Balance
£1890.00
1 unit
16354718
View accessory products View Documents
Sartorius
BCE423I-1CEU
420 g 1 mg CEU Electronic Toploading Balance
£2000.00
1 unit
16314738
View accessory products View Documents
Sartorius
BCE622I-1CEU
620 g 10 mg CEU Electronic Toploading Balance
£1685.00
1 unit
16354738
View accessory products View Documents
Sartorius
BCE623I-1CEU
620 g 1 mg CEU Electronic Toploading Balance
£2220.00
1 unit
16334748
View accessory products View Documents
Sartorius
BCE653I-1CEU
650 g 1 mg CEU Electronic Toploading Balance
£2435.00
1 unit
16354758
View accessory products View Documents
Sartorius
BCE822I-1CEU
820 g 10 mg CEU Electronic Toploading Balance
£1775.00
1 unit
16394678
View accessory products View Documents
Sartorius
BCE1202I-1CEU
1200 g 10 mg CEU Electronic Toploading Balance
£1890.00
1 unit
16374688
View accessory products View Documents
Sartorius
BCE2201I-1CEU
2200 g 100 mg CEU Electronic Toploading Balance
£1775.00
1 unit
16314698
View accessory products View Documents
Sartorius
BCE2202I-1CEU
2200 g 10 mg CEU Electronic Toploading Balance
£2000.00
1 unit
16334708
View accessory products View Documents
Sartorius
BCE3202I-1CEU
3200 g 10 mg CEU Electronic Toploading Balance
£2120.00
1 unit
16314718
View accessory products View Documents
Sartorius
BCE4202I-1CEU
4200 g 10 mg CEU Electronic Toploading Balance
£2220.00
1 unit
16394718
View accessory products View Documents
Sartorius
BCE5201I-1CEU
5200 g 100 mg CEU Electronic Toploading Balance
£1890.00
1 unit
16334728
View accessory products View Documents
Sartorius
BCE6200I-1CEU
6200 g 1000 mg CEU Electronic Toploading Balance
£1775.00
1 unit
16374728
View accessory products View Documents
Sartorius
BCE6202I-1CEU
6200 g 10 mg CEU Electronic Toploading Balance
£2435.00
1 unit
16374748
View accessory products View Documents
Sartorius
BCE8200I-1CEU
8200 g 1000 mg CEU Electronic Toploading Balance
£1890.00
1 unit
16314758
View accessory products View Documents
Sartorius
BCE8201I-1CEU
8200 g 100 mg CEU Electronic Toploading Balance
£2000.00
1 unit
Description
Description
  • No matter what you're weighing, the new Entris™ II balance is always the right choice.
  • Offering unrivalled value and backed by almost 150 years of German engineering expertise, the Entris™ II comes in two product lines, so you can find the balance that meets your specific weighing needs.
  • With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
Specifications
Specifications
CEU
Touchscreen
Precision Balance
RS232, USB C
PBT, Glass, Stainless Steel
100 – 240 VAC (±10%), 50 - 60 Hz, 0.2 A
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Entris™ II Precision Balance, Internal Adjustment