Learn More
Memmert™ Entry Port 14 mm, Flap Closure, Custom Position Tbc In The Back For S110
Optional entry port for cooled incubator. Memmert™ Entry Port, Pelitier-Cooled Incubator can be placed in special positions at the back (specify).
Brand: Memmert™ S110D6
Additional Details : Weight : 0.01100kg
Description
- Can be closed by flap
- In special positions on the back
- 14 mm clear diameter
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 | |
14 mm | |
Entry Port |