Learn More
Memmert™ Entry Port 14 mm, Flap Closure, Custom Position Tbc In The Back For S55
Optional entry port for cooled incubator. Memmert™ Entry Port, Pelitier-Cooled Incubator can be placed in special positions at the back (specify).
Brand:  Memmert™ S55D6
Additional Details : Weight : 0.01100kg
Product Code. 16395907
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Can be closed by flap
  • In special positions on the back
  • 14 mm clear diameter
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
14 mm
Entry Port