Learn More
Memmert™ Entry Port 14 mm for IN450plus
Optional entry port for Memmert Incubators.
Brand: Memmert™ IN450plusD6
Additional Details : Weight : 0.01200kg
Description
- Entry port with 14 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
IN450plus Incubator | |
Entry Port | |
14 mm |