Learn More
Memmert™ Entry Port 14 mm for UF260
Product Code. 17599125 Shop All Memmert Products
Click to view available options
Diameter (Metric):
14 mm
Unit Size:
Each
Product Code. 17599125
Brand: Memmert™ UF260D6
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UF260 Oven
Type Entry Port
Diameter (Metric) 14 mm