Learn More
Memmert™ Entry Port 23 mm, Flap Closure, Custom Position Tbc In The Back For S75
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand: Memmert™ S75F6
Additional Details : Weight : 0.01100kg
Description
- Entry port with clear diameter
- For Memmert Ovens
Specifications
Introducing Moisture Tight Connections, Can be Closed by Flap in Special Rear Positions | |
Entry Port | |
Stainless Steel |