missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port 23 mm, Flap & Stopper Closure, Standard Position Right Side Centre/Centre, For Un160Pa

Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.

Brand:  Memmert™ UN160paF2

Product Code. 16344357

  • £192.00 / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Entry port with clear diameter
  • For Memmert Ovens
Specifications

Specifications

Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Stainless Steel
23 mm
Entry Port
0.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Entry Port 23 mm, Flap & Stopper Closure, Standard Position Right Side Centre/Centre, For Un160Pa >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.