Learn More
Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For S75
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand:  Memmert™ S75F1
Additional Details : Weight : 0.01100kg
Product Code. 16376017
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Entry port with clear diameter
  • For Memmert Ovens
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
23 mm
Entry Port
Stainless Steel