Learn More
Memmert™ Entry Port 23 mm for UF30, Custom TBC Right Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF30F5
Product Code. 17599245
  • £166.00 / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in right side
Specifications
Specifications
UF30 Oven
Entry Port
23 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UF30, Custom TBC Right Side > For Use With: UF30