Learn More
Memmert™ Entry Port 23 mm for UF30, Standard Position Right Side Center/Center
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF30F2
Product Code. 17569245
  • £115.00 / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position right side center/center
Specifications
Specifications
UF30 Oven
Entry Port
23 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UF30, Standard Position Right Side Center/Center > For Use With: UF30