Learn More
Memmert™ Entry Port
Material: Silicone; For Use With: ICO240
Brand: Memmert™ ICO240F7
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Moisture tight
- Can be closed by silicone stopper
Specifications
Silicone | |
Accessory, Entry Port | |
40 mm |