Learn More
Memmert™ Entry Port Custom Position
Silicone Entry Port flap and stopper closure custom position TBC in the back
Brand: Memmert™ IPP1060ecoF7
Additional Details : Weight : 0.01100kg
Description
- Individual position at the back (please, state location; not in combination with illumination units)
- Moisture tight, can be closed by silicone stopper
Specifications
Clear | |
IPP1060eco | |
Silicone | |
40 mm |