Learn More
Memmert™ Entry Port Custom Position
Silicone Entry Port flap and stopper closure custom position TBC in the back
Brand:  Memmert™ HPP750ecoF7
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17139884
  • £353.66 / Each
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
  • Individual position at the back (please, state location; not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Specifications
Clear
HPP750eco
Silicone
40 mm