Learn More
Memmert™ Entry Port Left Centre/Centre
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
Brand: Memmert™ HPP260ecoF0
Additional Details : Weight : 0.01100kg
Description
- Standard position left centre/centre
- Not for model size 260 with light module
- Not for model size 110 with light module
Specifications
Moisture tight | |
HPP260eco |