Learn More
Memmert™ Entry Port Left Centre/Top
For introducing connections at the side and can be closed by flap
Brand: Memmert™ HPP260ecoF1
Additional Details : Weight : 0.01100kg
Description
- Standard position left centre/top
- Not for modelsize 110 with light module
Specifications
Clear | |
HPP260eco | |
23 mm |