Learn More
Memmert™ Entry Port Left (Individual Location)
Entry port flap closure custom TBC position in left side
Brand:  Memmert™ HPP2200ecoF4
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17189744
  • £231.47 / Each
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
  • Entry port flap closure custom TBC position in left side
  • Can be closed by the flap
Specifications
Specifications
Clear
Not in combination with illumination units, moisture tight, can be closed by flap and silicone stopper
HPP2200eco
23 mm