Learn More
Memmert™ Entry Port Rear (Individual Position)
Entry port with flap closure and custom position TBC in the back
Brand: Memmert™ HPP750ecoF6
Additional Details : Weight : 0.01100kg
Description
- Not in combination with illumination units
- Moisture tight
- Can be closed by flap and silicone stopper
Specifications
Clear | |
HPP750eco | |
23 mm |