Learn More
Memmert™ Entry Port Rear (Individual Position)
Entry port with flap closure and custom position TBC in the back
Brand:  Memmert™ IPP410ecoF6
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17130235
  • £155.75 / Each
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper
Specifications
Specifications
Clear
IPP410eco
23 mm