Learn More
Memmert™ Entry Port Right Centre/Centre
For introducing connections at the side and can be closed by flap
Brand: Memmert™ HPP110ecoF2
Additional Details : Weight : 0.01100kg
Description
- Standard position right centre/centre
- Not for model sizes 110 and 260 with light module
- Not for model size 110 with light module
Specifications
Clear | |
HPP110eco | |
23 mm |