Learn More
Memmert™ Entry Port Right Centre/Centre
For introducing connections at the side and can be closed by flap
Brand:  Memmert™ IPP110ecoF2
 View more versions of this product
Product Code. 17160105
  • £116.00 / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard position right centre/centre
  • Not for model sizes 110 and 260 with light module
  • Not for model size 110 with light module
Specifications
Specifications
Clear
IPP110eco
23 mm