Learn More
Memmert™ Entry Port Right (Individual Location)
Entry port with flap closure and custom TBC position in right side
Brand: Memmert™ HPP260ecoF5
Additional Details : Weight : 0.01100kg
Description
- Not in combination with illumination units
- Moisture tight
- Can be closed by flap and silicone stopper
Specifications
Clear | |
HPP260eco | |
23 mm |