Learn More
Memmert™ Entry Port Standard Position
Silicone Entry Port flap and stopper closure standard position in the back
Brand: Memmert™ IPP1400ecoD8
Additional Details : Weight : 0.01100kg
Description
- Standard position rear (not in combination with illumination units)
- Moisture tight, can be closed by silicone stopper
Specifications
Clear | |
IPP1400eco | |
Silicone | |
40 mm |