Learn More
Memmert™ Entry Port Standard Position
Silicone Entry Port flap and stopper closure standard position in the back
Brand:  Memmert™ IPP750ecoD8
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17140265
  • £274.50 / Each
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
  • Standard position rear (not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Specifications
Clear
IPP750eco
Silicone
40 mm