Learn More
Cytiva (formerly Pall Lab) Envirochek™ Polyester HV Sampling Capsule
Envirochek™ HV capsules are approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts.
Brand:  Cytiva (formerly Pall Lab) 12099
Additional Details : Weight : 0.22700kg
 View more versions of this product
Product Code. 17174221
  • £196.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
  • Approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts from up to 50 L of water according to EPA method 1623.1, with a faster, safer, and simpler protocol than other methods including processing of up to 8 samples at once using our shaker
Specifications
Specifications
Urethane Adhesive
1 μm
1300 cm2
1/Ea.
Spun Polypropylene
Non-sterile
Polycarbonate
Straight Hose Barb