Learn More
Neogen Environmental Collection Swab ECS100
Product Code. 17283814
Encompass
Click to view available options
Unit Size:
Pack of 100
Product Code. 17283814
Brand: Neogen 7100177463
The sterile, individually wrapped polyester swabs for environmental sampling in the food and beverage industry are intended for use with Neogen Allergen Test Kits
  • Sterilized swab in individual packaging
  • Swab for use with Neogen Allergenic Protein Test Kits
  • 100 /Pk.
Specifications
Sterility Sterile
Packaging 100 Pk.
For Use With (Application) 3 M™ Allergenic Protein Test Kits
Product Type Swab