missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Caspase-3 Activity Assay Kits

Detect apoptosis in cell extracts and purified enzyme preparations with EnzChek Caspase-3 Activity Assay Kits with Z-DEVD-AMC substrate or Z-DEVD-R110 substrates.

£594.00 - £642.00

Specifications

Quantity 500 assays
Format Cuvets, 96-well Plate
Product Type Caspase Assay Kit
No. of Reactions 500 assays (reaction volume 100μL per assay)
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Description Label or Dye Price Quantity & Availability  
Product Code Brand Description Label or Dye Price Quantity & Availability  
10174472
View Documents
Invitrogen™
E13183
EnzChek Caspase-3 Assay Kit #1, Z-DEVD-AMC substrate AMC (7-amino-4-methylcoumarin)
£594.00
Each
Please to purchase this item. Need a web account? Register with us today!
10573473
View Documents
Invitrogen™
E13184
EnzChek Caspase-3 Assay Kit #2, Z-DEVD-R110 substrate Rhodamine 110
£642.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Enable simple and reliable detection of caspase-3-activity-mediated apoptosis with the EnzChek Caspase-3 Activity Assay Kit, which takes advantage of specially designed substrates that fluoresce upon cleavage by the caspase-3 enzyme. The EnzChek Caspase-3 Activity Assay Kit is available with either the Z-DEVD-AMC or Z-DEVD-R110 substrate, which yields a bright fluorescent product upon proteolytic cleavage by caspase 3/7.

The EnzChek Caspase-3 Activity Assay Kit enables detection of apoptosis by providing a simple and reliable method for assaying caspase-3/7 activity. The kit can be used to continuously measure the activity of caspase-3/7 in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader.

Key features of the EnzChek Caspase-3 Activity Assay Kit includes:
• Fluorescent assay using standard fluorescein (FITC) excitation/emission settings
• Format allows for continuous measurement of caspase-3/7 activity in cell extracts
• Fluorescent and enzymatic controls included

The aminomethylcoumarin (AMC)-derived substrate (Z-aspartic acid-glutamic acid-valine-aspartic acid (DEVD)-AMC) that is included in the EnzChek Caspase-3 Activity Assay Kit is weakly fluorescent in the UV range (excitation/emission maxima ˜330/390 nm), but yields a bright, blue-fluorescent product (excitation/emission maxima ˜342/441 nm) upon proteolytic cleavage. The kit can be used to continuously measure the activity of caspase-3 and closely related proteases in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader.

The rhodamine 110-derived substrate (Z-DEVD-R110) used in the EnzChek Caspase-3 Activity Assay Kit is a non-fluorescent bisamide compound that, upon enzymatic cleavage, is converted in a two-step process to the fluorescent monoamide and then to the even more fluorescent R110 product. Both of these hydrolysis products exhibit spectral properties similar to those of fluorescein, with peak excitation and emission wavelengths of 496 nm and 520 nm, respectively.

In addition to the substrates, the EnzChek Caspase Activity Assay Kit contains the reversible aldehyde inhibitor Ac-DEVD-CHO, as well as an AMC or R110 reference standard. The Ac-DEVD-CHO inhibitor confirms that the fluorescence signals in both induced and control cell populations are due to the activity of the caspase-3/7 protease. The reference standard allows for quantification of the amount of AMC or R110 released in the reaction.
Specifications

Specifications

500 assays
Caspase Assay Kit
Room Temperature
Fluorescence
Other Label(s) or Dye(s)
Cuvets, 96-well Plate
500 assays (reaction volume 100μL per assay)
EnzChek
Fluorometer, Microplate Reader
Store in freezer (-5 to -30°C) and protect from light.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.