missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin B, pure, high purity, biological stain

Product Code. 10174660
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Eosin B, C20H6Br2N2Na2O9, CAS Number-548-24-3, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, Eosin Bluish, 100g, 0.96 to 1.22, 19, 233, Brown, 208-943-1, 624.06, Pure, Danger, GHS H Statement: Causes serious eye damage.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
MDL Number MFCD00005041
Synonym 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish

Specifications

Absorption Ratio 0.96 to 1.22
Beilstein 19, 233
Merck Index 15, 3657
Infrared Spectrum Authentic
Lambda Max 511 to 520nm (in water + 1% Na2CO3)
Formula Weight 624.06
Grade Pure
Packaging Glass bottle
Color Brown
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Eosin B
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.