missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Eosin Y, pure, high purity, biological stain
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | >300.0°C |
Dye Content | 90% min. (on dry substance) |
Beilstein | 19, IV, 2917 |
Merck Index | 15, 3658 |
Solubility Information | Solubility in water: freely soluble. Other solubilities: sparingly soluble in alcohol, insoluble in diethyl ether |
Infrared Spectrum | Authentic |
Formula Weight | 691.85 |
Loss on Drying | 10% max. |
Grade | Pure |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Eosin Y
- Warning
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Eosin Y
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction