missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, high purity, biological stain

Product Code. 10676584
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2', 4', 5'
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Melting Point >300.0°C
Dye Content 90% min. (on dry substance)
Beilstein 19, IV, 2917
Merck Index 15, 3658
Solubility Information Solubility in water: freely soluble. Other solubilities: sparingly soluble in alcohol, insoluble in diethyl ether
Infrared Spectrum Authentic
Formula Weight 691.85
Loss on Drying 10% max.
Grade Pure
Packaging Glass bottle
Specific Gravity 1.018
Density 1.0180g/mL
Color Brown to Red
Quantity 500 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Eosin Y, High purity biological stain
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.