missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CCL24 (Eotaxin-2) Recombinant Protein

Eotaxin-2 Recombinant Mouse Protein

Brand:  Gibco™ PMC0144

Product Code. 10263113

  • £387.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Recombinant mouse eotaxin is a bioactive protein intended for use in cell culture applications. Eotaxin is a member of the PF-4 family of chemokines. Eotaxin plays a role in inducing calcium flux in eosinophils.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mammalian Cell Culture
20292
Eotaxin-2 Recombinant Mouse Protein
20 μg
For research use only. Not for use in diagnostic procedures
Eotaxin-2
The biological activity was determined by measuring dose dependent chemotaxis with murine lymphocytes. A concentration range of 10 to 100ng/mL produced significant chemotaxis. Eotaxin-2, a CC chemokine, is characterized by its high chemotactic selectivity for eosinophils. The receptor for eotaxin-2 is CCR3
Mouse
Chemokines and receptors
Eotaxins
Lyophilized
10.3kDa
Sequential chromatography
2 to 8°C
1 Each
Eotaxin-2 Recombinant Mouse Protein
Eotaxin-2 Recombinant Mouse Protein
E. coli
Lyophilized
>98% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.