Learn More
PeproTech Eotaxin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P67BT-500UG
Description
AA Sequence of recombinant protein: HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine Eotaxin (CCL11). Anti-Murine Eotaxin (CCL11)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mEotaxin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine Eotaxin (500-P67) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mEotaxin. Western Blot: To detect mEotaxin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mEotaxin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P67BT-1MG will be provided as 2 x 500 μg
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
| Eotaxin | |
| Polyclonal | |
| Biotin | |
| CCL11 | |
| C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20292 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P48298 | |
| CCL11 | |
| E.coli-derived Recombinant Murine Eotaxin (CCL11). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.