missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epa Method 8081 Pesticide Standard, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Description
200μg/mL, 1mL
Specifications
Quantity | 1 mL |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- PAH MIX A
- Danger
- Acute toxicity Category 2
- Aspiration toxicity Category 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Carcinogenicity Category 1A
- Reproductive toxicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Germ cell mutagenicity Category 1B
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H372-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dichloromethane, benzene, anthracene, carbazole, naphthalene, chrysene
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'