Learn More
Fisherbrand™ EPA Vial Kit, 24-400 screw thread, Clear Glass, White PP screw cap, Center hole, Silicone/PTFE septum
Includes 100 vials and 100 caps with pre-assembled septa.
Brand:  Fisherbrand™ 24 14 1278
135.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12980991
  • £149.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • ND24
  • 24-400
  • Glass
  • Clear
  • Screw Cap
  • Center hole
  • Polypropylene
  • Silicone/PTFE
  • Reusable two compartment trays protect vials and closure while keeping matching supplies together
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White Silicone/Beige PTFE
27.5 mm
60 mL
24-400
Screw Cap
1st Hydrolytic Class
Clear
Glass
TOC (EPA)
Polypropylene
140 mm