Learn More
BRAND™ EPDM Rubber Conical Gaskets
Place between filter funnel and filter flask for a secure fit.
Brand:  BRAND™ 463139
119.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10277971
  • £137.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
71 x 52 mm dia.
10
71 mm
Rubber Conical Gasket
Used for Filter Funnels and Filter Flasks
71 mm
EPDM