Learn More
BRAND™ EPDM Rubber Conical Gaskets
Place between filter funnel and filter flask for a secure fit.
Brand:  BRAND™ 462500
 View more versions of this product
Product Code. 10614552
  • £73.15 / Pack of 8
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22 to 84 mm dia.
8
22 to 84 mm
Rubber Gasket Set
Used to Place between Filter Funnel and Filter Flask
22 - 84 mm
EPDM