missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13724-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. The protein encoded by this gene functions as a receptor for ephrin A2, A3 and A5 and plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.Specifications
EPHA8 | |
Polyclonal | |
Unconjugated | |
EPHA8 | |
EEK, EK3, EPH and ELK related kinase, EPH like kinase 3, EPH receptor A8, EPHA8, Ephrin type A receptor 8, HEK3, KIAA1459 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13842, 2046, 60589 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O09127, P29321, P29322 | |
Epha8 | |
EPHA8 Fusion Protein Ag4503 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |