missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin, 99%

Product Code. p-4509556 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
This item is not returnable. View return policy

Product Code. 10071080

Brand: Thermo Scientific Acros 117780250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 106-89-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl

Spezifikation

Melting Point -57.0°C
Density 1.1800g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4385
Quantity 25 mL
Beilstein 17,6
Fieser 01,355; 04,222; 05,290
Merck Index 15,3666
Specific Gravity 1.18
Solubility Information Solubility in water: 60g/l (10°C). Other solubilities: misclible with alcohol, ether, chloroform,, trichloroethylene, carbon tetrachloride, acetone, and benzene
Viscosity 1.12 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Epichlorohydrin
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Epichlorohydrin
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt