missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ EpiLife™ Medium, with 60 μM calcium
EpiLife™ Medium, with 60 μM calcium
Brand: Gibco™ MEPI500CA
77.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01050kg
Description
A sterile, liquid medium prepared with 60μM calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of an appropriate growth supplement prior to use. EpiLife™ greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife™ medium is HEPES and bicarbonate buffered, and is designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Liquid | |
Serum-free | |
Sterile-filtered | |
Sustainable packaging | |
Cascade Biologics™, EpiLife™ |
Keratinocyte Cells, Epithelial Cells | |
Mammalian Cell Culture | |
EpiLife Medium | |
Sterile-filtered | |
HEPES, Phenol Red | |
Calcium | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.