missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%, Thermo Scientific Chemicals

£81.50 - £356.00

Chemical Identifiers

CAS 3221-15-6
Molecular Formula C3H5ClS
Molecular Weight (g/mol) 108.59
MDL Number MFCD00041199
InChI Key XRWMHJJHPQTTLQ-UHFFFAOYSA-N
Synonym 2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
PubChem CID 18588
IUPAC Name 2-(chloromethyl)thiirane
SMILES C1C(S1)CCl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10033490
View Documents
Thermo Scientific Acros
291390050
5 g Glass bottle
£81.50
5g
Please to purchase this item. Need a web account? Register with us today!
10628074
View Documents
Thermo Scientific Acros
291390250
25 g Glass bottle
£356.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3221-15-6
108.59
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
18588
C1C(S1)CCl
C3H5ClS
MFCD00041199
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
2-(chloromethyl)thiirane

Specifications

3221-15-6
100.0
1.2750g/mL
Authentic
C3H5ClS
MFCD00041199
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
2-(chloromethyl)thiirane
18588
97%
Epithiochlorohydrine
96.0
Colorless to Yellow
69°C to 70°C (67.0 mmHg)
96% min. (GC)
1.5240 to 1.528
1.275
Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
C1C(S1)CCl
108.59
108.59
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

Product Identifier
  • Epithiochlorohydrine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only