Learn More
Epo Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA185404
Code : LD
Additional Details : Weight : 0.01000kg
Description
Rabbit anti Human erythropoietin antibody reacts with erythropoietin, the principal hormone involved in the regulation of erythrocyte differentiation and the maintenance of a physiological levels of circulating erythrocyte mass. It is often used as a drug to treat anemia.
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.Specifications
Epo | |
Polyclonal | |
Unconjugated | |
EPO | |
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
Rabbit | |
Protein A | |
RUO | |
2056 | |
-20°C | |
Liquid |
ELISA, Functional Assay, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with <5% trehalose and no preservative | |
P01588 | |
EPO | |
CHO cell-derived, recombinant human erythropoietin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |