missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Epoch 2 Microplate Spectrophotometer
GREENER_CHOICE

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.

POA

Specifications

Detector Type Photodiode
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Accuracy ±2 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
View More Specs

Products 4
Product Code Brand Model Includes Price Quantity & Availability  
Product Code Brand Model Includes Price Quantity & Availability  
17227809
GREENER_CHOICE
missing translation for 'viewDocuments'
Agilent BioTek
EPOCH2NS-SN
EPOCH2NS No Touchscreen; No cuvette port
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17247809
GREENER_CHOICE
missing translation for 'viewDocuments'
Agilent BioTek
EPOCH2NSC-SN
EPOCH2NSC Cuvette Port
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17267809
GREENER_CHOICE
missing translation for 'viewDocuments'
Agilent BioTek
EPOCH2TS-SN
EPOCH2TS Touchscreen only
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17287809
GREENER_CHOICE
missing translation for 'viewDocuments'
Agilent BioTek
EPOCH2TSC-SN
EPOCH2TSC Touchscreen; Cuvette port
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths

Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.

Specifications

Specifications

Photodiode
96 wells: 8 sec., 384 wells: 14 sec.
CE and TUV marked. RoHS compliant.
22 in.
200 to 999 nm, 1 nm increments
Xenon Flash Lamp
±2 nm
Nucleic acid and protein quantification to ELISA and microbial growth assays
50/60 Hz
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Agilent BioTek Epoch 2 Microplate Spectrophotometer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.