Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
POA
Specifications
Detector Type | Photodiode |
---|---|
Light Source | Xenon Flash Lamp |
Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
Accuracy | ±2 nm |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
17257809
![]() |
Agilent BioTek
EPOCH2TS-SI |
EPOCH2TS | Touchscreen only |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17277809
![]() |
Agilent BioTek
EPOCH2TSC-SI |
EPOCH2TSC | Touchscreen; Cuvette port |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17217809
![]() |
Agilent BioTek
EPOCH2NS-SI |
EPOCH2NS | No Touchscreen; No cuvette port |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17237809
![]() |
Agilent BioTek
EPOCH2NSC-SI |
EPOCH2NSC | Cuvet port only |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Specifications
Photodiode | |
96 wells: 8 sec., 384 wells: 14 sec. | |
CE and TUV marked. RoHS compliant. | |
22 in. | |
200 to 999 nm, 1 nm increments |
Xenon Flash Lamp | |
±2 nm | |
Nucleic acid and protein quantification to ELISA and microbial growth assays | |
50/60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.