missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epoprostenol, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
Descripción
Endogenous prostanoid that is a potent agonist at IP prostanoid receptors. Inhibits platelet aggregation and induces vasodilation.
Especificaciones
Especificaciones
CAS | 61849-14-7 |
Molecular Formula | C20H31NaO5 |
Quantity | 10 mg |
Synonym | (5Z,13E,8R,9S,11R,12R,15S)-6,9-Epoxy-11,15-dihydroxyprosta-5,13-dien-1-oic acid sodium salt |
InChI Key | LMHIPJMTZHDKEW-HIHHSTLZSA-M |
SMILES | CCCCCC(C=CC1C(CC2C1CC(=CCCCC(=O)[O-])O2)O)O.[Na+] |
IUPAC Name | sodium;(5Z)-5-[(3aS,4S,5S,6aR)-5-hydroxy-4-[(E,3R)-3-hydroxyoct-1-enyl]-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-ylidene]pentanoate |
Molecular Weight (g/mol) | 374.453 |
PubChem CID | 133687047 |
Formula Weight | 374.45 |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'