Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Brand:  Eppendorf™ 0030133374
147.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10297987
  • £155.00 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
Standard
Autoclavable
Colorless
Unskirted, Divisible
250 μL
Non-sterile
2250 x g
PCR Clean (Non-sterile)
Colorless
Human DNA, DNase, RNase-free
8 x 12
20 Plates (5 Bags x 4 Plates)
PCR Plate
Polypropylene, Polycarbonate
96
Ideal for extreme applications such as multiple heat sealing.
Regular
20 Pk.