missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips
GREENER_CHOICE

Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.

Brand:  Eppendorf™ 0030089715

9.75 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.


 View more versions of this product

Product Code. 12614607

  • £11.09 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.

  • Available in 25 and 50mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
1 Piece
Red
25 mL Eppendorf™ Quality Tip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers