Learn More
Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids |
Tip Style | Low Retention |
Description
- Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
- Tips are uncoated and free from additives
- Ultra-homogenous surface for maximum reproducibility
- Ultra-hydrophobic surface for minimum loss of sample
- Air bubbles and foaming are significantly reduced during pipetting
- Autoclavable, high chemical resistance
- Five-year shelf life
Specifications
Autoclavable | |
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids | |
Low Retention |